Assay Details
Target Gene Details
Entrez Gene ID: | 3604 |
Gene Name: | TNF receptor superfamily member 9 |
Gene Aliases: |
4-1BB, CD137, CDw137, ILA |
Location: |
Chr.1:7915871-7943165 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF9 | NM_001561.5 | 9 | 5464 | NP_001552.2 |
| XM_011541386.2 | 8 | 5479 | XP_011539688.1 | |
| AW449076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2678721 | Chr.1:7915332 - 7918244 on Build GRCh38 | Deletion |
|
| esv3585133 | Chr.1:7915221 - 7918142 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map