Assay Details
Target Gene Details
Entrez Gene ID: | 2046 |
Gene Name: | EPH receptor A8 |
Gene Aliases: |
EEK, EK3, HEK3 |
Location: |
Chr.1:22563508-22603594 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA8 | NM_001006943.1 | NP_001006944.1 | ||
| XM_011540969.1 | XP_011539271.1 | |||
| XM_011540970.1 | XP_011539272.1 | |||
| XM_011540972.1 | XP_011539274.1 | |||
| BC038796.1 | AAH38796.1 | |||
| BC072417.1 | ||||
| DR007053.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26050 | Chr.1:22562682 - 22564686 on Build GRCh38 | Gain |
|
| dgv183n54 | Chr.1:22563356 - 22564576 on Build GRCh38 | Loss |
|
| dgv184n54 | Chr.1:22563554 - 22564322 on Build GRCh38 | Loss |
|
| nsv545750 | Chr.1:22559273 - 22564428 on Build GRCh38 | Loss |
|
| nsv509024 | Chr.1:22536990 - 22581743 on Build GRCh38 | Insertion |
|
| nsv545753 | Chr.1:22562979 - 22564162 on Build GRCh38 | Loss |
|
| dgv185n54 | Chr.1:22563949 - 22564630 on Build GRCh38 | Gain |
|
| dgv186n54 | Chr.1:22563965 - 22564422 on Build GRCh38 | Loss |
|
| nsv545749 | Chr.1:22551320 - 22625893 on Build GRCh38 | Loss |
|
| dgv181n54 | Chr.1:22562710 - 22564576 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map