Assay Details
Target Gene Details
Entrez Gene ID: | 1952 |
Gene Name: | cadherin EGF LAG seven-pass G-type receptor 2 |
Gene Aliases: |
ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3 |
Location: |
Chr.1:109249705-109275756 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELSR2 | NM_001408.2 | NP_001399.1 | ||
| XM_005270580.4 | XP_005270637.1 | |||
| AF234887.1 | AAG00080.1 | |||
| D87469.2 | BAA13407.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv993086 | Chr.1:109254248 - 109263222 on Build GRCh38 | Deletion |
|
| nsv470732 | Chr.1:109239415 - 109275908 on Build GRCh38 | Loss |
|
| nsv1012267 | Chr.1:109168205 - 109279887 on Build GRCh38 | Gain |
|
| nsv951381 | Chr.1:109248679 - 109277078 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map