Assay Details
Target Gene Details
Entrez Gene ID: | 192669 |
Gene Name: | argonaute 3, RISC catalytic component |
Gene Aliases: |
EIF2C3 |
Location: |
Chr.1:35925198-36072037 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGO3 | NM_024852.3 | NP_079128.2 | ||
| NM_177422.2 | NP_803171.1 | |||
| XM_005270575.3 | XP_005270632.1 | |||
| XM_011540879.2 | XP_011539181.1 | |||
| XM_017000523.1 | XP_016856012.1 | |||
| AK022827.1 | BAB14262.1 | |||
| AK027796.1 | BAB55374.1 | |||
| AK292870.1 | ||||
| BC066888.1 | AAH66888.1 | |||
| BF438817.1 | ||||
| BF979526.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115413630] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map