Assay Details
Target Gene Details
Entrez Gene ID: | 84251 |
Gene Name: | SH3 domain GRB2 like endophilin interacting protein 1 |
Gene Aliases: |
- |
Location: |
Chr.1:66533272-66751139 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGIP1 | NM_001308203.1 | NP_001295132.1 | ||
| XM_005271270.4 | 1 | 135 | XP_005271327.1 | |
| XM_017002506.1 | 1 | 144 | XP_016857995.1 | |
| XM_017002508.1 | 1 | 145 | XP_016857997.1 | |
| XM_017002510.1 | 1 | 144 | XP_016857999.1 | |
| XM_017002511.1 | 1 | 144 | XP_016858000.1 | |
| XM_017002512.1 | XP_016858001.1 | |||
| XM_017002516.1 | 1 | 144 | XP_016858005.1 | |
| XM_017002517.1 | 1 | 143 | XP_016858006.1 | |
| XM_017002518.1 | 1 | 143 | XP_016858007.1 | |
| XM_017002522.1 | 1 | 143 | XP_016858011.1 | |
| XM_017002524.1 | 1 | 137 | XP_016858013.1 | |
| XM_017002525.1 | 1 | 136 | XP_016858014.1 | |
| XM_017002527.1 | XP_016858016.1 | |||
| XM_017002528.1 | 1 | 137 | XP_016858017.1 | |
| XM_017002533.1 | 1 | 135 | XP_016858022.1 | |
| XM_017002535.1 | 1 | 135 | XP_016858024.1 | |
| XM_017002537.1 | 1 | 134 | XP_016858026.1 | |
| BC040516.1 | AAH40516.1 | |||
| BX640813.1 | CAE45891.1 | |||
| CR749541.1 | CAH18344.1 | |||
| DA724991.1 |
More Information
Set Membership: |
|

Genomic Map