Assay Details
Target Gene Details
Entrez Gene ID: | 642946 |
Gene Name: | FLVCR1 antisense RNA 1 (head to head) |
Gene Aliases: |
LQK1, NCRNA00292 |
Location: |
Chr.1:212856604-212858138 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLVCR1-AS1 | NR_027285.1 | 2 | 588 | |
| NR_027286.1 | ||||
| AK092887.1 | ||||
| AY030238.1 | 2 | 527 | AAK51141.1 | |
| AY030239.1 | AAK51142.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv572n100 | Chr.1:212856472 - 212899671 on Build GRCh38 | Gain |
|
| dgv571n100 | Chr.1:212831823 - 212896221 on Build GRCh38 | Gain |
|
| esv3891115 | Chr.1:212828746 - 212857815 on Build GRCh38 | Loss |
|
| nsv1008383 | Chr.1:212852204 - 212941552 on Build GRCh38 | Gain |
|

Genomic Map