Assay Details
Target Gene Details
Entrez Gene ID: | 8289 |
Gene Name: | AT-rich interaction domain 1A |
Gene Aliases: |
B120, BAF250, BAF250a, BM029, C1orf4, CSS2, ELD, MRD14, OSA1, P270, SMARCF1, hELD, hOSA1 |
Location: |
Chr.1:26696031-26782110 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID1A | NM_006015.4 | NP_006006.3 | ||
| NM_139135.2 | NP_624361.1 | |||
| AB210022.1 | BAE06104.1 | |||
| AF219114.1 | AAG17549.2 | |||
| AF231056.1 | AAG33967.1 | |||
| AF265208.1 | AAF75765.1 | |||
| AF521670.1 | AAN03446.1 |
More Information
Set Membership: |
|

Genomic Map