Assay Details
Target Gene Details
Entrez Gene ID: | 127845 |
Gene Name: | golgi transport 1A |
Gene Aliases: |
CGI-141, GOT1, HMFN1187, YMR292W |
Location: |
Chr.1:204198158-204214092 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLT1A | NM_198447.1 | NP_940849.1 | ||
| XM_017000314.1 | XP_016855803.1 | |||
| AB075871.1 | BAD38653.1 | |||
| AK124645.1 | BAC85915.1 | |||
| BC058832.1 | AAH58832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762138 | Chr.1:204138959 - 204248018 on Build GRCh38 | Gain |
|
| nsv549039 | Chr.1:204192463 - 204305128 on Build GRCh38 | Gain |
|
| nsv1007228 | Chr.1:204174999 - 204216444 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map