Assay Details
Target Gene Details
Entrez Gene ID: | 100874253 |
Gene Name: | long intergenic non-protein coding RNA 853 |
Gene Aliases: |
PDZK1IP1-AS1, ncRNA-a4 |
Location: |
Chr.1:47179250-47180339 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00853 | NR_047498.1 | |||
| AI792909.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950709 | Chr.1:47173129 - 47191428 on Build GRCh38 | Deletion |
|

Genomic Map