Assay Details
Target Gene Details
Entrez Gene ID: | 6894 |
Gene Name: | TAR (HIV-1) RNA binding protein 1 |
Gene Aliases: |
TRM3, TRP-185, TRP185 |
Location: |
Chr.1:234391313-234479174 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TARBP1 | NM_005646.3 | NP_005637.3 | ||
| XM_005273234.3 | XP_005273291.1 | |||
| XM_011544263.2 | XP_011542565.1 | |||
| XM_017002195.1 | XP_016857684.1 | |||
| XM_017002196.1 | XP_016857685.1 | |||
| XM_017002197.1 | XP_016857686.1 | |||
| U38847.1 | AAC50379.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv585n100 | Chr.1:234254426 - 234483126 on Build GRCh38 | Gain |
|
| esv3589170 | Chr.1:234394180 - 234413960 on Build GRCh38 | Loss |
|
| esv3589169 | Chr.1:234392482 - 234485360 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map