Assay Details
Target Gene Details
Entrez Gene ID: | 22823 |
Gene Name: | metal response element binding transcription factor 2 |
Gene Aliases: |
M96, PCL2, TDRD19A, dJ976O13.2 |
Location: |
Chr.1:93079219-93139081 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTF2 | NM_001164391.1 | NP_001157863.1 | ||
| NM_001164392.1 | NP_001157864.1 | |||
| NM_001164393.1 | NP_001157865.1 | |||
| NM_007358.3 | NP_031384.1 | |||
| XM_011541016.1 | XP_011539318.1 | |||
| XM_011541017.2 | XP_011539319.1 | |||
| XM_011541019.1 | XP_011539321.1 | |||
| XM_017000676.1 | XP_016856165.1 | |||
| XM_017000677.1 | XP_016856166.1 | |||
| XM_017000678.1 | XP_016856167.1 | |||
| XM_017000679.1 | XP_016856168.1 | |||
| AF072814.1 | AAC27618.1 | |||
| AF086908.1 | AAP97138.1 | |||
| AJ010014.1 | CAA08970.1 | |||
| AK290318.1 | ||||
| AK302776.1 | ||||
| AK302900.1 | ||||
| BC010013.2 | AAH10013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830625 | Chr.1:93100821 - 93269689 on Build GRCh38 | Loss |
|
| nsv1008602 | Chr.1:92711254 - 93175251 on Build GRCh38 | Gain |
|
| nsv508404 | Chr.1:93081826 - 93114455 on Build GRCh38 | Deletion |
|
| nsv830614 | Chr.1:93000421 - 93213143 on Build GRCh38 | Loss |
|
| nsv1866 | Chr.1:93095373 - 93127336 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map