Assay Details
Target Gene Details
Entrez Gene ID: | 10327 |
Gene Name: | aldo-keto reductase family 1 member A1 |
Gene Aliases: |
ALDR1, ALR, ARM, DD3, HEL-S-6 |
Location: |
Chr.1:45550779-45570051 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR1A1 | NM_001202413.1 | NP_001189342.1 | ||
| NM_001202414.1 | NP_001189343.1 | |||
| NM_006066.3 | NP_006057.1 | |||
| NM_153326.2 | NP_697021.1 | |||
| XM_011540491.2 | XP_011538793.1 | |||
| XM_011540492.2 | XP_011538794.1 | |||
| AK293083.1 | ||||
| AK307068.1 | ||||
| AK307237.1 | ||||
| AK307288.1 | ||||
| AK311175.1 | ||||
| AU142853.1 | ||||
| BC000670.2 | AAH00670.1 | |||
| BC005394.1 | AAH05394.1 | |||
| BI549149.1 | ||||
| CX788871.1 | ||||
| DA550374.1 | ||||
| DA871469.1 | ||||
| EU668319.1 | ||||
| EU794588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain |
|
| nsv829726 | Chr.1:45431909 - 45637514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map