Assay Details
Target Gene Details
Entrez Gene ID: | 22996 |
Gene Name: | tetratricopeptide repeat domain 39A |
Gene Aliases: |
C1orf34, DEME-6 |
Location: |
Chr.1:51287258-51345116 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC39A | NM_001144832.2 | NP_001138304.1 | ||
| NM_001297662.1 | NP_001284591.1 | |||
| NM_001297663.1 | NP_001284592.1 | |||
| NM_001297664.1 | NP_001284593.1 | |||
| XM_005270644.2 | XP_005270701.1 | |||
| XM_006710471.2 | XP_006710534.1 | |||
| XM_011541048.2 | XP_011539350.1 | |||
| XM_011541050.2 | XP_011539352.1 | |||
| XM_011541051.2 | XP_011539353.1 | |||
| XM_011541053.2 | XP_011539355.1 | |||
| XM_017000715.1 | XP_016856204.1 | |||
| AB007921.1 | BAA32297.1 | |||
| AF007170.1 | AAC39582.1 | |||
| AK000220.1 | ||||
| AK298019.1 | ||||
| AK301587.1 | ||||
| AK311430.1 | ||||
| BI761577.1 | ||||
| CB128766.1 | ||||
| CX788693.1 | ||||
| DA871604.1 | ||||
| DC381029.1 |
Target Gene Details
Entrez Gene ID: | 102724097 |
Gene Name: | TTC39A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:51329654-51331281 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC39A-AS1 | NR_123731.1 | |||
| BF675965.1 | ||||
| CB128747.1 |
More Information
Set Membership: |
|

Genomic Map