Assay Details
Target Gene Details
Entrez Gene ID: | 100506007 |
Gene Name: | RTCA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:100264742-100266174 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTCA-AS1 | NR_110434.1 | 3 | 513 | |
| NR_110435.1 | 3 | 499 | ||
| DA390082.1 | 3 | 499 | ||
| DA695476.1 | 3 | 464 |

Genomic Map