Assay Details
Target Gene Details
Entrez Gene ID: | 23139 |
Gene Name: | microtubule associated serine/threonine kinase 2 |
Gene Aliases: |
MAST205, MTSSK |
Location: |
Chr.1:45786992-46036124 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAST2 | NM_001319245.1 | NP_001306174.1 | ||
| NM_001324320.1 | NP_001311249.1 | |||
| NM_015112.2 | NP_055927.2 | |||
| XM_011541059.2 | XP_011539361.1 | |||
| XM_011541061.2 | XP_011539363.1 | |||
| XM_011541062.2 | XP_011539364.1 | |||
| XM_011541063.1 | XP_011539365.1 | |||
| XM_011541064.2 | XP_011539366.1 | |||
| XM_011541065.1 | XP_011539367.1 | |||
| XM_011541069.2 | XP_011539371.1 | |||
| AB047005.1 | BAB40778.1 | |||
| BC065499.1 | AAH65499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585914 | Chr.1:45718616 - 45877574 on Build GRCh38 | Gain |
|
| nsv1014128 | Chr.1:45631998 - 45928868 on Build GRCh38 | Gain |
|
| nsv461417 | Chr.1:45757414 - 45950676 on Build GRCh38 | Gain |
|
| esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain |
|
| nsv524147 | Chr.1:45770426 - 45972404 on Build GRCh38 | Gain |
|
| dgv159n100 | Chr.1:45706164 - 45997340 on Build GRCh38 | Gain |
|
| nsv1013628 | Chr.1:45755945 - 45890893 on Build GRCh38 | Gain |
|
| nsv546158 | Chr.1:45750903 - 46029762 on Build GRCh38 | Gain |
|
| esv3585915 | Chr.1:45720361 - 46036324 on Build GRCh38 | Gain |
|
| esv3577360 | Chr.1:45654763 - 45881711 on Build GRCh38 | Gain |
|
| nsv437479 | Chr.1:45807207 - 45945474 on Build GRCh38 | Loss |
|
| nsv1009527 | Chr.1:45820657 - 46054455 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map