Assay Details
Target Gene Details
Entrez Gene ID: | 57172 |
Gene Name: | calcium/calmodulin dependent protein kinase IG |
Gene Aliases: |
CLICK3, CLICKIII, VWS1, dJ272L16.1 |
Location: |
Chr.1:209583700-209613939 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMK1G | NM_020439.2 | NP_065172.1 | ||
| XM_017001866.1 | XP_016857355.1 | |||
| XM_017001867.1 | XP_016857356.1 | |||
| AF428261.1 | AAL28100.1 | |||
| AK095713.1 | ||||
| AK289675.1 | ||||
| BC032787.1 | AAH32787.1 | |||
| BG715920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3588705 | Chr.1:209586152 - 209587922 on Build GRCh38 | Loss |
|
| esv2672471 | Chr.1:209586219 - 209587914 on Build GRCh38 | Deletion |
|
| esv3575639 | Chr.1:209579620 - 209597199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map