Assay Details
Target Gene Details
Entrez Gene ID: | 6900 |
Gene Name: | contactin 2 |
Gene Aliases: |
AXT, FAME5, TAG-1, TAX, TAX1 |
Location: |
Chr.1:205043160-205078043 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTN2 | NM_005076.3 | NP_005067.1 | ||
| XM_017002199.1 | XP_016857688.1 | |||
| CN289415.1 | ||||
| CR749495.1 | CAH18318.1 | |||
| X67734.1 | CAA47963.1 | |||
| X68274.1 | CAA48335.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549041 | Chr.1:205006607 - 205072337 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map