Assay Details
Target Gene Details
Entrez Gene ID: | 83479 |
Gene Name: | DEAD-box helicase 59 |
Gene Aliases: |
OFD5, ZNHIT5 |
Location: |
Chr.1:200640807-200669998 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX59 | NM_001320181.1 | 8 | 1977 | NP_001307110.1 |
| XM_006711562.1 | XP_006711625.1 | |||
| AK299219.1 | 8 | 1848 | ||
| AL136611.1 | CAB66546.1 | |||
| BM683465.1 | 1 | 331 |
More Information
Set Membership: |
|

Genomic Map