Assay Details
Target Gene Details
Entrez Gene ID: | 100506696 |
Gene Name: | prostate cancer associated transcript 6 (non-protein coding) |
Gene Aliases: |
KDM5B-AS1, KDM5BAS1, PCAN-R1, ncRNA-a2, onco-lncRNA-96 |
Location: |
Chr.1:202810946-202811913 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCAT6 | NR_046325.1 | 2 | 688 | |
| NR_046326.1 | 2 | 574 | ||
| AI806248.1 | 1 | 81 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832348 | Chr.1:202641288 - 202813034 on Build GRCh38 | Loss |
|
| nsv4110 | Chr.1:202787512 - 202820558 on Build GRCh38 | Insertion |
|
| nsv521757 | Chr.1:202811402 - 202812036 on Build GRCh38 | Loss |
|
| nsv832359 | Chr.1:202739288 - 202918866 on Build GRCh38 | Loss |
|

Genomic Map