Assay Details
Target Gene Details
Entrez Gene ID: | 23032 |
Gene Name: | ubiquitin specific peptidase 33 |
Gene Aliases: |
VDU1 |
Location: |
Chr.1:77695987-77759879 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP33 | NM_015017.4 | NP_055832.3 | ||
| NM_201624.2 | NP_963918.1 | |||
| XM_005270648.1 | XP_005270705.1 | |||
| XM_005270649.1 | XP_005270706.1 | |||
| XM_011541055.1 | XP_011539357.1 | |||
| XM_017000722.1 | XP_016856211.1 | |||
| XM_017000723.1 | XP_016856212.1 | |||
| XM_017000724.1 | XP_016856213.1 | |||
| XM_017000725.1 | XP_016856214.1 | |||
| XM_017000726.1 | XP_016856215.1 | |||
| XM_017000727.1 | XP_016856216.1 | |||
| XM_017000728.1 | XP_016856217.1 | |||
| XM_017000729.1 | XP_016856218.1 | |||
| AB029020.1 | ||||
| AF383172.1 | AAL78314.1 | |||
| AF383173.1 | AAL78315.1 | |||
| AK023664.1 | ||||
| AL050140.1 | CAB43288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470719 | Chr.1:77308728 - 77803522 on Build GRCh38 | Gain |
|
| nsv830303 | Chr.1:77625139 - 77843080 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114025509] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map