Assay Details
Target Gene Details
Entrez Gene ID: | 51097 |
Gene Name: | saccharopine dehydrogenase (putative) |
Gene Aliases: |
CGI-49, NET11 |
Location: |
Chr.1:246724076-246768137 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCCPDH | NM_016002.2 | NP_057086.2 | ||
| AF151807.1 | AAD34044.1 | |||
| AK075178.1 | BAC11453.1 | |||
| BC026185.2 | AAH26185.1 | |||
| BG258610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2667587 | Chr.1:246652751 - 246920518 on Build GRCh38 | Deletion |
|
| esv2762192 | Chr.1:246660555 - 246905930 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map