Assay Details
Target Gene Details
Entrez Gene ID: | 51253 |
Gene Name: | mitochondrial ribosomal protein L37 |
Gene Aliases: |
L37mt, MRP-L2, MRP-L37, MRPL2, RPML2 |
Location: |
Chr.1:54200167-54225489 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL37 | NM_016491.3 | NP_057575.2 | ||
| XM_005270920.3 | XP_005270977.1 | |||
| AF151069.1 | AAF36155.1 | |||
| AF325707.1 | AAG52881.1 | |||
| AK024572.1 | ||||
| AK026171.1 | ||||
| AY421759.1 | AAR38839.1 | |||
| BC000041.2 | AAH00041.1 |
More Information
Set Membership: |
|

Genomic Map