Assay Details
Target Gene Details
Entrez Gene ID: | 1677 |
Gene Name: | DNA fragmentation factor subunit beta |
Gene Aliases: |
CAD, CPAN, DFF-40, DFF2, DFF40 |
Location: |
Chr.1:3857143-3885429 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DFFB | NM_001282669.1 | NP_001269598.1 | ||
| NM_001320132.1 | NP_001307061.1 | |||
| NM_001320136.1 | NP_001307065.1 | |||
| NM_004402.3 | NP_004393.1 | |||
| NR_104222.1 | ||||
| NR_135150.1 | ||||
| NR_135151.1 | ||||
| NR_135152.1 | ||||
| XM_017000498.1 | XP_016855987.1 | |||
| XM_017000499.1 | XP_016855988.1 | |||
| XM_017000500.1 | XP_016855989.1 | |||
| AB013918.1 | BAA32250.1 | |||
| AB028911.1 | BAB40447.1 | |||
| AB028912.1 | BAB40448.1 | |||
| AB028913.1 | BAB40449.1 | |||
| AF039210.1 | AAC39709.1 | |||
| AF064019.1 | AAC39920.1 | |||
| AF409060.1 | AAL02005.1 | |||
| AF409061.1 | AAL02006.1 | |||
| AF409062.1 | AAL02007.1 | |||
| AK290877.1 | ||||
| AK303065.1 | ||||
| AY438267.1 | AAR05821.1 | |||
| AY438268.1 | AAR05822.1 | |||
| BC032827.1 | AAH32827.1 | |||
| BC048797.1 | AAH48797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain |
|
| esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain |
|
| nsv509590 | Chr.1:3826457 - 3902492 on Build GRCh38 | Insertion |
|
| esv3585052 | Chr.1:3817921 - 3928224 on Build GRCh38 | Gain |
|
| nsv545130 | Chr.1:3826568 - 4283888 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72848499] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map