Assay Details
Target Gene Details
Entrez Gene ID: | 2046 |
Gene Name: | EPH receptor A8 |
Gene Aliases: |
EEK, EK3, HEK3 |
Location: |
Chr.1:22563508-22603594 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA8 | NM_001006943.1 | NP_001006944.1 | ||
| XM_011540969.1 | XP_011539271.1 | |||
| XM_011540970.1 | XP_011539272.1 | |||
| XM_011540972.1 | XP_011539274.1 | |||
| XM_011540973.1 | XP_011539275.1 | |||
| BC038796.1 | AAH38796.1 | |||
| BC072417.1 | ||||
| DR007053.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509024 | Chr.1:22536990 - 22581743 on Build GRCh38 | Insertion |
|
| esv994542 | Chr.1:22574893 - 22580151 on Build GRCh38 | Insertion |
|
| nsv545749 | Chr.1:22551320 - 22625893 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map