Assay Details
Target Gene Details
Entrez Gene ID: | 5051 |
Gene Name: | platelet activating factor acetylhydrolase 2 |
Gene Aliases: |
HSD-PLA2 |
Location: |
Chr.1:25959764-25998164 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAFAH2 | NM_000437.3 | 11 | 2871 | NP_000428.2 |
| XM_006710669.3 | 8 | 3163 | XP_006710732.1 | |
| XM_006710670.3 | 11 | 2936 | XP_006710733.1 | |
| XM_011541528.2 | 12 | 3002 | XP_011539830.1 | |
| XM_011541529.2 | 9 | 2624 | XP_011539831.1 | |
| XM_017001390.1 | 11 | 2897 | XP_016856879.1 | |
| XM_017001391.1 | 10 | 2672 | XP_016856880.1 | |
| XM_017001392.1 | 10 | 2743 | XP_016856881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834502 | Chr.1:25834281 - 25987428 on Build GRCh38 | Gain |
|
| nsv834513 | Chr.1:25922980 - 26087385 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map