Assay Details
Target Gene Details
Entrez Gene ID: | 26750 |
Gene Name: | ribosomal protein S6 kinase C1 |
Gene Aliases: |
RPK118, RSKL1, S6K-delta-1, S6PKh1, humS6PKh1 |
Location: |
Chr.1:213051233-213378753 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Additional Information:
For this assay, SNP(s) [rs74362592] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map