Assay Details
Target Gene Details
Entrez Gene ID: | 10911 |
Gene Name: | urotensin 2 |
Gene Aliases: |
PRO1068, U-II, UCN2, UII |
Location: |
Chr.1:7846345-7913615 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UTS2 | XM_011540537.2 | XP_011538839.1 | ||
| XM_017000116.1 | XP_016855605.1 | |||
| XM_017000119.1 | XP_016855608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3567897 | Chr.1:7861892 - 7899931 on Build GRCh38 | Gain |
|
| esv3585132 | Chr.1:7861408 - 7892708 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72633060] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map