Assay Details
Target Gene Details
Entrez Gene ID: | 6491 |
Gene Name: | SCL/TAL1 interrupting locus |
Gene Aliases: |
MCPH7, SIL |
Location: |
Chr.1:47250139-47314787 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160846 | Chr.1:47249998 - 47266706 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map