Assay Details
Target Gene Details
Entrez Gene ID: | 2052 |
Gene Name: | epoxide hydrolase 1 |
Gene Aliases: |
EPHX, EPOX, HYL1, MEH |
Location: |
Chr.1:225810074-225845563 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHX1 | NM_000120.3 | NP_000111.1 | ||
| NM_001136018.3 | NP_001129490.1 | |||
| NM_001291163.1 | NP_001278092.1 | |||
| AK313436.1 | ||||
| BC003567.1 | AAH03567.1 | |||
| BC008291.1 | AAH08291.1 | |||
| BC095430.1 | AAH95430.1 | |||
| CR541963.1 | CAG46761.1 | |||
| J03518.1 | AAA61305.1 | |||
| KF032815.1 | ||||
| KU178042.1 | ||||
| KU178043.1 | ||||
| L25878.1 | AAA52389.1 | |||
| L25879.1 | AAA52390.1 | |||
| M36374.1 | AAA59580.1 | |||
| X07936.1 | CAA30759.1 | |||
| Y00424.1 | CAA68486.1 |
Target Gene Details
Entrez Gene ID: | 9725 |
Gene Name: | transmembrane protein 63A |
Gene Aliases: |
KIAA0792 |
Location: |
Chr.1:225840556-225882720 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM63A |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762223 | Chr.1:225837474 - 226014015 on Build GRCh38 | Gain |
|
| nsv524126 | Chr.1:225786075 - 226022952 on Build GRCh38 | Gain |
|
| nsv520628 | Chr.1:225838705 - 225905662 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map