Assay Details
Target Gene Details
Entrez Gene ID: | 3694 |
Gene Name: | integrin subunit beta 6 |
Gene Aliases: |
AI1H |
Location: |
Chr.2:160099666-160200313 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB6 | NM_000888.4 | 15 | 4306 | NP_000879.2 |
| NM_001282353.1 | 16 | 4199 | NP_001269282.1 | |
| NM_001282354.1 | 13 | 4021 | NP_001269283.1 | |
| NM_001282355.1 | 13 | 3985 | NP_001269284.1 | |
| NM_001282388.1 | 14 | 4226 | NP_001269317.1 | |
| NM_001282389.1 | 15 | 4133 | NP_001269318.1 | |
| NM_001282390.1 | 14 | 4053 | NP_001269319.1 | |
| AI264144.1 | 1 | 388 | ||
| AK026736.1 | 1 | 1701 | ||
| AL359053.1 | 1 | 1447 | ||
| AL831998.1 | 15 | 4245 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010774 | Chr.2:159012247 - 160175703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map