Assay Details
Target Gene Details
Entrez Gene ID: | 23671 |
Gene Name: | transmembrane protein with EGF like and two follistatin like domains 2 |
Gene Aliases: |
CT120.2, HPP1, TENB2, TPEF, TR, TR-2 |
Location: |
Chr.2:191948299-192194940 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEFF2 | NM_001305134.1 | NP_001292063.1 | ||
| NM_016192.3 | NP_057276.2 | |||
| XM_011510890.2 | XP_011509192.1 | |||
| XM_017003739.1 | XP_016859228.1 | |||
| AB017269.1 | BAA87897.1 | |||
| AF179274.2 | AAD55776.2 | |||
| AF242222.1 | AAG49452.1 | |||
| AK074507.1 | BAC11030.1 | |||
| AK074632.1 | ||||
| AK222691.1 | BAD96411.1 | |||
| AK315376.1 | ||||
| AL157430.1 | CAB75654.1 | |||
| AY358907.1 | AAQ89266.1 | |||
| BC008973.1 | AAH08973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584068 | Chr.2:191838120 - 192003002 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map