Assay Details
Target Gene Details
Entrez Gene ID: | 57628 |
Gene Name: | dipeptidyl peptidase like 10 |
Gene Aliases: |
DPL2, DPPY, DPRP-3, DPRP3 |
Location: |
Chr.2:114442358-115845752 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPP10 | NM_001178036.1 | NP_001171507.1 | ||
| NM_001321905.1 | NP_001308834.1 | |||
| NM_001321907.1 | NP_001308836.1 | |||
| NM_001321908.1 | NP_001308837.1 | |||
| NM_001321910.1 | NP_001308839.1 | |||
| NM_001321911.1 | NP_001308840.1 | |||
| NM_001321912.1 | NP_001308841.1 | |||
| NM_001321913.1 | NP_001308842.1 | |||
| NM_020868.4 | NP_065919.2 | |||
| AK055256.1 | ||||
| AK289967.1 | ||||
| AK315147.1 | ||||
| AY172661.3 | AAO17263.2 | |||
| BC030832.1 | AAH30832.1 |
Target Gene Details
Entrez Gene ID: | 101927591 |
Gene Name: | DPP10 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.2:114828341-114833548 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPP10-AS3 | NR_132105.1 | |||
| HY010208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893148 | Chr.2:114765597 - 115116964 on Build GRCh38 | Gain |
|
| nsv459019 | Chr.2:114737565 - 114841436 on Build GRCh38 | Loss |
|
| esv3893137 | Chr.2:114489425 - 116378222 on Build GRCh38 | Gain |
|
| esv3592057 | Chr.2:114794993 - 114837510 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map