Assay Details
Target Gene Details
Entrez Gene ID: | 6869 |
Gene Name: | tachykinin receptor 1 |
Gene Aliases: |
NK1R, NKIR, SPR, TAC1R |
Location: |
Chr.2:75046463-75199519 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TACR1 | NM_001058.3 | 5 | 2318 | NP_001049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998145 | Chr.2:74969670 - 75241030 on Build GRCh38 | Loss |
|
| nsv834262 | Chr.2:74907892 - 75062072 on Build GRCh38 | Gain |
|
| nsv1001495 | Chr.2:74598036 - 75093218 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map