Assay Details
Target Gene Details
Entrez Gene ID: | 151393 |
Gene Name: | regulator of microtubule dynamics 2 |
Gene Aliases: |
BLOCK18, FAM82A, FAM82A1, PRO34163, PYST9371, RMD-2, RMD2, RMD4 |
Location: |
Chr.2:37920789-38068032 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMDN2 | XM_011532617.2 | XP_011530919.1 | ||
| XM_011532618.2 | XP_011530920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6787n54 | Chr.2:37855471 - 37965002 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map