Assay Details
Target Gene Details
Entrez Gene ID: | 6519 |
Gene Name: | solute carrier family 3 member 1 |
Gene Aliases: |
ATR1, CSNU1, D2H, NBAT, RBAT |
Location: |
Chr.2:44275458-44320824 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC3A1 | NM_000341.3 | NP_000332.2 | ||
| XM_011533047.2 | XP_011531349.1 | |||
| AB033549.1 | BAB16841.1 | |||
| AK091932.1 | ||||
| AK092000.1 | ||||
| AK223146.1 | BAD96866.1 | |||
| AK289636.1 | ||||
| AK309145.1 | ||||
| AK313157.1 | ||||
| BC022386.1 | AAH22386.1 | |||
| BC093624.1 | AAH93624.1 | |||
| BC093626.1 | AAH93626.1 | |||
| DQ023512.1 | AAY89643.1 | |||
| DQ023513.1 | AAY89644.1 | |||
| DQ023515.1 | AAY89646.1 | |||
| DQ023516.1 | AAY89647.1 | |||
| DQ023517.1 | AAY89648.1 | |||
| KU178335.1 | ||||
| KU178336.1 | ||||
| L11696.1 | AAA81778.1 | |||
| M95548.1 | AAA35500.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3798n100 | Chr.2:44279994 - 44404126 on Build GRCh38 | Loss |
|
| nsv581692 | Chr.2:44278046 - 44322729 on Build GRCh38 | Loss |
|
| esv3590554 | Chr.2:44290134 - 44306750 on Build GRCh38 | Gain |
|
| esv3575237 | Chr.2:44290145 - 44318453 on Build GRCh38 | Gain |
|
| dgv3799n100 | Chr.2:44284605 - 44325240 on Build GRCh38 | Loss |
|
| nsv1006405 | Chr.2:44013164 - 44709137 on Build GRCh38 | Loss |
|
| nsv1011655 | Chr.2:44278899 - 44344334 on Build GRCh38 | Loss |
|
| nsv581691 | Chr.2:44276990 - 44339733 on Build GRCh38 | Gain |
|
| nsv521399 | Chr.2:44280074 - 44317775 on Build GRCh38 | Loss |
|
| nsv581695 | Chr.2:44292003 - 44318437 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map