Assay Details
Target Gene Details
Entrez Gene ID: | 1796 |
Gene Name: | docking protein 1 |
Gene Aliases: |
P62DOK, pp62 |
Location: |
Chr.2:74549020-74557551 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOK1 | NM_001197260.1 | NP_001184189.1 | ||
| NM_001318866.1 | 1 | 341 | NP_001305795.1 | |
| NM_001318867.1 | 1 | 341 | NP_001305796.1 | |
| NM_001318868.1 | 1 | 341 | NP_001305797.1 | |
| NM_001318869.1 | 1 | 341 | NP_001305798.1 | |
| NM_001381.4 | 1 | 341 | NP_001372.1 | |
| AF035299.1 | AAB88182.1 | |||
| AK055944.1 | 1 | 341 | ||
| DA382785.1 | ||||
| DA599419.1 | 1 | 113 |
Target Gene Details
Entrez Gene ID: | 84695 |
Gene Name: | lysyl oxidase like 3 |
Gene Aliases: |
LOXL |
Location: |
Chr.2:74532258-74555709 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL3 | XM_011533134.2 | XP_011531436.1 |
More Information
Set Membership: |
|

Genomic Map