Assay Details
Target Gene Details
Entrez Gene ID: | 101928161 |
Gene Name: | uncharacterized LOC101928161 |
Gene Aliases: |
- |
Location: |
Chr.2:133266195-133284758 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928161 | NR_110294.1 | |||
| DB498836.1 |
Target Gene Details
Entrez Gene ID: | 344148 |
Gene Name: | NCK associated protein 5 |
Gene Aliases: |
ERIH1, ERIH2, NAP5 |
Location: |
Chr.2:132671788-133675181 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCKAP5 | NM_207363.2 | NP_997246.2 | ||
| NM_207481.3 | NP_997364.3 | |||
| XM_005263659.3 | XP_005263716.1 | |||
| XM_005263660.3 | XP_005263717.1 | |||
| XM_011511097.2 | XP_011509399.1 | |||
| XM_011511098.2 | XP_011509400.1 | |||
| XM_011511099.2 | XP_011509401.1 | |||
| XM_011511100.2 | XP_011509402.1 | |||
| XM_011511101.1 | XP_011509403.1 | |||
| XM_011511102.2 | XP_011509404.1 | |||
| XM_011511104.1 | XP_011509406.1 | |||
| XM_017003974.1 | XP_016859463.1 | |||
| XM_017003975.1 | XP_016859464.1 | |||
| XM_017003976.1 | XP_016859465.1 | |||
| XM_017003977.1 | XP_016859466.1 | |||
| XM_017003979.1 | XP_016859468.1 | |||
| AK057980.1 | ||||
| AK092189.1 | BAC03824.1 | |||
| BC110831.1 | AAI10832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516915 | Chr.2:133242259 - 133297090 on Build GRCh38 | Gain |
|
| nsv1011100 | Chr.2:133142490 - 133288591 on Build GRCh38 | Loss |
|
| nsv583152 | Chr.2:132369502 - 133631550 on Build GRCh38 | Gain |
|
| nsv1003964 | Chr.2:133281791 - 133330747 on Build GRCh38 | Loss |
|
| nsv834384 | Chr.2:133247657 - 133431077 on Build GRCh38 | Gain |
|
| esv3893403 | Chr.2:133198640 - 133353352 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map