Assay Details
Target Gene Details
Entrez Gene ID: | 140739 |
Gene Name: | ubiquitin conjugating enzyme E2 F (putative) |
Gene Aliases: |
NCE2 |
Location: |
Chr.2:237966945-238042782 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2F | NM_001278305.1 | NP_001265234.1 | ||
| NM_001278306.1 | NP_001265235.1 | |||
| NM_001278307.1 | NP_001265236.1 | |||
| NM_001278308.1 | NP_001265237.1 | |||
| NM_080678.2 | NP_542409.1 | |||
| NR_103498.1 | ||||
| NR_103499.1 | ||||
| NR_103500.1 | ||||
| AF310723.1 | AAL26792.1 | |||
| AK290063.1 | ||||
| AK293334.1 | ||||
| AK294107.1 | ||||
| AK297502.1 | ||||
| AK303094.1 | ||||
| BC010549.1 | AAH10549.1 | |||
| BM920749.1 | ||||
| BQ959565.1 | ||||
| BX421077.2 | ||||
| DA140546.1 | ||||
| DB118122.1 |
Target Gene Details
Entrez Gene ID: | 100533179 |
Gene Name: | UBE2F-SCLY readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.2:237966945-238099413 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2F-SCLY | NR_037904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4193n100 | Chr.2:237988669 - 238064519 on Build GRCh38 | Gain |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map