Assay Details
Target Gene Details
Entrez Gene ID: | 117581 |
Gene Name: | twist family bHLH transcription factor 2 |
Gene Aliases: |
AMS, BBRSAY, DERMO1, FFDD3, SETLSS, bHLHa39 |
Location: |
Chr.2:238848032-238910548 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TWIST2 | NM_001271893.3 | NP_001258822.1 | ||
| AW072674.1 | ||||
| BC017907.1 | AAH17907.1 | |||
| BC103755.1 | AAI03756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3594773 | Chr.2:238869973 - 238892363 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74000306] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map