Assay Details
Target Gene Details
Entrez Gene ID: | 79172 |
Gene Name: | centromere protein O |
Gene Aliases: |
CENP-O, ICEN-36, MCM21R |
Location: |
Chr.2:24793115-24822376 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPO | NM_001199803.2 | NP_001186732.1 | ||
| NM_001322101.1 | NP_001309030.1 | |||
| NM_024322.3 | NP_077298.1 | |||
| NR_136182.1 | ||||
| NR_136183.1 | ||||
| NR_136184.1 | ||||
| AK027859.1 | BAB55414.1 | |||
| AK315483.1 | ||||
| AL514456.3 | ||||
| BC002870.1 | AAH02870.1 | |||
| BP366489.1 | ||||
| BX406097.2 | ||||
| CF993857.1 | ||||
| CN263048.1 | ||||
| CX872478.1 | ||||
| DA001951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004529 | Chr.2:24363242 - 25005468 on Build GRCh38 | Loss |
|
| nsv508831 | Chr.2:24787088 - 24857561 on Build GRCh38 | Insertion |
|
| nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain |
|
| nsv1000020 | Chr.2:24709475 - 24909903 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map