Assay Details
Target Gene Details
Entrez Gene ID: | 9736 |
Gene Name: | ubiquitin specific peptidase 34 |
Gene Aliases: |
- |
Location: |
Chr.2:61187455-61470714 on Build GRCh38 |
Assay Gene Location: | Within Intron 68 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP34 | NM_014709.3 | NP_055524.3 | ||
| AB011142.2 | BAA25496.2 | |||
| AB018272.1 | BAA34449.1 | |||
| AJ586138.1 | CAE51938.1 | |||
| AK001193.1 | ||||
| AK023845.1 | ||||
| AK125898.1 | ||||
| AL050092.1 | CAB43264.1 | |||
| BC022783.1 | AAH22783.1 |
More Information
Set Membership: |
|

Genomic Map