Assay Details
Target Gene Details
Entrez Gene ID: | 8291 |
Gene Name: | dysferlin |
Gene Aliases: |
FER1L1, LGMD2B, MMD1 |
Location: |
Chr.2:71453155-71686763 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582197 | Chr.2:71480078 - 71489364 on Build GRCh38 | Loss |
|
| nsv525538 | Chr.2:71484819 - 71488316 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map