Assay Details
Target Gene Details
Entrez Gene ID: | 56171 |
Gene Name: | dynein axonemal heavy chain 7 |
Gene Aliases: |
- |
Location: |
Chr.2:195737604-196075188 on Build GRCh38 |
Assay Gene Location: | Within Intron 61 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAH7 | NM_018897.2 | NP_061720.2 | ||
| XM_011511487.2 | XP_011509789.1 | |||
| XM_011511488.2 | XP_011509790.1 | |||
| XM_011511489.2 | XP_011509791.1 | |||
| XM_011511490.2 | XP_011509792.1 | |||
| XM_011511497.2 | XP_011509799.1 | |||
| XM_017004504.1 | XP_016859993.1 | |||
| AB023161.2 | ||||
| AF327442.1 | AAL37427.1 | |||
| BC029567.1 | AAH29567.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76920418] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map