Assay Details
Target Gene Details
Entrez Gene ID: | 84695 |
Gene Name: | lysyl oxidase like 3 |
Gene Aliases: |
LOXL |
Location: |
Chr.2:74532258-74555709 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL3 | NM_001289164.1 | NP_001276093.1 | ||
| NM_001289165.1 | NP_001276094.1 | |||
| NM_032603.3 | NP_115992.1 | |||
| XM_011533134.2 | XP_011531436.1 | |||
| XM_017005112.1 | XP_016860601.1 | |||
| AF282619.1 | AAK51671.1 | |||
| AF284815.1 | AAK91134.1 | |||
| AF311313.1 | AAK63205.1 | |||
| AK309474.1 | ||||
| AK309478.1 | ||||
| BC071865.1 | AAH71865.1 | |||
| DQ378059.1 | ABD23013.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs80153151] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map