Assay Details
Target Gene Details
Entrez Gene ID: | 56888 |
Gene Name: | potassium channel modulatory factor 1 |
Gene Aliases: |
DEBT91, FIGC, PCMF, ZZZ1 |
Location: |
Chr.2:84971108-85059472 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCMF1 | NM_020122.4 | NP_064507.3 | ||
| XM_006712052.3 | XP_006712115.1 | |||
| XM_011532990.1 | XP_011531292.1 | |||
| AB083199.1 | BAC43745.1 | |||
| AF155652.1 | AAF67009.1 | |||
| AK023403.1 | BAB14563.1 | |||
| AK314761.1 | ||||
| BC000178.2 | AAH00178.1 | |||
| BU844982.1 | ||||
| CT004343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834279 | Chr.2:84980330 - 85178181 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114127529] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map