Assay Details
Target Gene Details
Entrez Gene ID: | 105373805 |
Gene Name: | uncharacterized LOC105373805 |
Gene Aliases: |
- |
Location: |
Chr.2:191021526-191032314 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373805 | NR_136318.1 | |||
| AW873644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2622574 | Chr.2:191023919 - 191025444 on Build GRCh38 | Deletion |
|
| nsv999391 | Chr.2:190884309 - 191057205 on Build GRCh38 | Gain |
|
| nsv834493 | Chr.2:190946647 - 191109514 on Build GRCh38 | Gain |
|

Genomic Map