Assay Details
Target Gene Details
Entrez Gene ID: | 4705 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A10 |
Gene Aliases: |
CI-42KD, CI-42k |
Location: |
Chr.2:239892450-240025402 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA10 | NR_136158.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997288 | Chr.2:239542278 - 240534009 on Build GRCh38 | Gain |
|
| nsv1006293 | Chr.2:239566358 - 239913428 on Build GRCh38 | Gain |
|
| nsv834584 | Chr.2:239892475 - 240037701 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76494079] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map