Assay Details
Target Gene Details
Entrez Gene ID: | 25797 |
Gene Name: | glutaminyl-peptide cyclotransferase |
Gene Aliases: |
GCT, QC, sQC |
Location: |
Chr.2:37344610-37373322 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QPCT | NM_012413.3 | NP_036545.1 | ||
| AK290605.1 | ||||
| BC036721.1 | AAH36721.1 | |||
| BC047756.2 | AAH47756.1 | |||
| BI755122.1 | ||||
| X67731.1 | CAA47961.1 | |||
| X71125.1 | CAA50438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2686 | Chr.2:37347524 - 37380543 on Build GRCh38 | Insertion |
|
| nsv2685 | Chr.2:37322953 - 37367780 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76199342] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map