Assay Details
Target Gene Details
Entrez Gene ID: | 55289 |
Gene Name: | acyl-CoA oxidase-like |
Gene Aliases: |
- |
Location: |
Chr.2:110732539-111118571 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv730e214 | Chr.2:110395308 - 110848620 on Build GRCh38 | Gain |
|
| nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss |
|
| nsv582699 | Chr.2:110763166 - 112172487 on Build GRCh38 | Loss |
|
| dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss |
|
| esv3591965 | Chr.2:110642083 - 110829253 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map