Assay Details
Target Gene Details
Entrez Gene ID: | 50940 |
Gene Name: | phosphodiesterase 11A |
Gene Aliases: |
PPNAD2 |
Location: |
Chr.2:177623249-178108339 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE11A | NM_001077196.1 | 17 | 2986 | NP_001070664.1 |
| NM_001077197.1 | 21 | 3516 | NP_001070665.1 | |
| NM_016953.3 | 20 | 4485 | NP_058649.3 | |
| AB036704.1 | BAB16371.1 | |||
| AB038041.1 | BAB16372.1 | |||
| AK074273.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583689 | Chr.2:177603280 - 177645092 on Build GRCh38 | Loss |
|
| esv3593466 | Chr.2:177618127 - 177839958 on Build GRCh38 | Gain |
|
| nsv583688 | Chr.2:177562912 - 177675565 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map